Learn More
Epredia™ Giemsa Stain
Take advantage of an excellent classical blood film stain used on peripheral blood smears and bone marrow with the Epredia™ Giemsa Stain.
460.00€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12583687
|
Epredia™
89004 |
1 gal. | N/A | ||||||
Description
- Erythrocytes stain pink with platelets exhibiting a very light, pale hue
- Leukocyte nuclear chromatin stains magenta-purple
- Monocyte cytoplasm stains the classical “ground glass” blue
- Lymphocyte cytoplasm stains clear, sky blue
Specifications
Giemsa Stain |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- STAIN,GIEMSA,1GL CUBE
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H330-Fatal if inhaled.
- H370-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P320-Specific treatment is urgent (see on this label).
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : Methanol