Learn More
Alla France™ General Purpose Density Hydrometer
Brand:  Alla France™ 3100MC170/20/H-qp
37.85 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11793423
  • 57.90€ / Each
Only null left
Estimated Shipment: 21-10-2025
Specifications
1Each
Density Hydrometer
250 mm
0.170 to 0.180 g/mL