Learn More
Alla France™ General Purpose Density Hydrometer
Brand:  Alla France™ 3100MC160/20/H-qp
37.75 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11783423
  • 62.70€ / Each
Only null left
Estimated Shipment: 17-09-2025
Specifications
1Each
Density Hydrometer
250 mm
0.160 to 0.170 g/mL