Learn More
Alla France™ General Purpose Density Hydrometer
Brand:  Alla France™ 3100MC060/20/B-qp
30.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11783413
  • 46.60€ / Each
Only null left
Estimated Shipment: 15-09-2025
Specifications
1Each
Density Hydrometer
250 mm
0.600 to 0.700 g/mL