Learn More
Alla France™ General Purpose Density Hydrometer
Brand:  Alla France™ 3100MC150/20/H-qp
30.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11773423
  • 50.35€ / Each
Only null left
Estimated Shipment: 12-09-2025
Specifications
1Each
Density Hydrometer
250 mm
0.150 to 0.160 g/mL