Learn More
BINDER™ Gas-tight Option Model
For Use With: FD 56
Brand: BINDER™ 8012-1683
1329.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Includes additional measures for the greatest possible impermeability to gas
- Not in conjunction with access port or interior lighting option
Specifications
Gas-tight option model - Incl. additional measures for the greatest possible impermeability to gas; not in conjunction with access port or interior lighting option | |
BINDER Chambers FD 56 | |
Steel | |
Gas-tight Option Model |