Learn More
Spex SamplePrep™ Freezer/Mill™ Dual Chamber Cryogenic Grinder
High-throughput, dual chamber cryogenic grinder with self-contained liquid nitrogen tub and insulated case.
Brand:  Spex SamplePrep™ 6875D-230
24508.63 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15595546
  • 32160.00€ / Each
Only null left
Estimated Shipment: 12-09-2025
Description
  • Designed for cryogenic grinding of tough and temperature sensitive samples
  • Mill cools samples to cryogenic temperatures
  • Liquid nitrogen required for operation
Specifications
Specifications
Freezer/Mill Dual Chamber Cryogenic Grinder
50.8 x 53.34 x 64.77 cm