Learn More
Fisherbrand™ Flask Clamp for Shaking Incubators
Fisherbrand™ Flask Clamp is designed to be used with a shaking incubator and can hold a variety of flask sizes.
Brand: Fisherbrand™ H1000-MR-CMB
57.54 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Constructed of stainless steel
- For use with shaking incubators
- Designed to hold flasks
Specifications
125, 250, 500 mL Flasks | |
Stainless Steel | |
1 Ea. |
Flask Clamp | |
Mini Shaking Incubator |