Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Alumina Neutral
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
414.00€ - 2470.00€
Specifications
Material | Polypropylene |
---|---|
Type | Neutral Alumina |
Resolution | 50 to 75 μm |
Column Type | Alumina Neutral |
For Use With (Application) | Polar Compounds |
Product Code | Brand | Size (Metric) Tube Rack | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) Tube Rack | Includes | Price | Quantity & Availability | ||||
16380216
|
BUCHI
140000103 |
1600 g | 3 Pc. |
2065.00€
Pack of 3 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||
16340216
|
BUCHI
140000099 |
160 g | 10 Pc. |
774.00€
Pack of 10 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||
16350216
|
BUCHI
140000100 |
240 g | 10 Pc. |
1275.00€
Pack of 10 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||
16310216
|
BUCHI
140000096 |
24 g | 18 Pc. |
414.00€
Pack of 18 |
In Stock
Log in to see stock. ![]() |
||||
16390216
|
BUCHI
140000104 |
3200 g | 2 Pc. |
2470.00€
Pack of 2 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||
16360216
|
BUCHI
140000101 |
440 g | 6 Pc. |
1120.00€
Pack of 6 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||
16320216
|
BUCHI
140000097 |
50 g | 12 Pc. |
456.00€
Pack of 12 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
16370216
|
BUCHI
140000102 |
660 g | 5 Pc. |
1490.00€
Pack of 5 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||
16330216
|
BUCHI
140000098 |
80 g | 12 Pc. |
544.00€
Pack of 12 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
Description
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
Polypropylene | |
50 to 75 μm | |
Polar Compounds |
Neutral Alumina | |
Alumina Neutral | |
Any Flash System |