Learn More
BINDER™ Shelf, Reinforced
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system
Brand:  BINDER™ 8012-0818
511.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15661916
  • 565.00€ / Each
Only null left
Estimated Shipment: 18-09-2025
Description
  • Reinforced
  • Positioned at bottom level
  • Maximum load of 45 kg
Specifications
Specifications
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system
Max. load of 67 kg
Reinforced Shelf
BINDER Chambers KT 53
Steel