Learn More
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks
High resistance to heat, chemical attack, and mechanical shock
Specifications
| Description | Top style: Heavy duty tooled with capacity scale |
|---|---|
| Material | Borosilicate Glass |
| No. of Positions | 1 |
| Writing Surface | Matte Finish Ceramic Enamel Marking |
| Disposable | No |
Description
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks offer sturdy borosilicate glass construction with a heavy-duty, tooled-top finish. High resistance to heat, chemical attack, and mechanical shock.
- Capacity scale measures approximate volume
- Matte-finish ceramic marking spot takes writing and erases easily
Compliance
- Designed from ASTM™ Specification E1404, Type I, Class 2 requirementsr
Specifications
| Top style: Heavy duty tooled with capacity scale | |
| 1 | |
| No |
| Borosilicate Glass | |
| Matte Finish Ceramic Enamel Marking | |
| Wide |