Learn More
Fisherbrand™ Polypropylene Graduated Container Lids
For polypropylene graduated containers 14-955-101,14-955-103, 14-955-104 and 14-955-106
Brand: Fisherbrand™ 14955108
25.12 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
For use with the following Fisherbrand Specimen Containers:
- 14-955-101, -103 (4.5 oz.)
- 14-955-104, -106 (8 oz.)
Specifications
Polyethylene | |
Not Autoclavable | |
500/Pk. |
Non-sterile | |
Polypropylene graduated containers |