Learn More
Fisherbrand™ Erlenmeyer Flask Clamps for Ultrasonic Units
Safely and securely attach Erlenmeyer flasks to Fisherbrand™ ultrasonic cleaning units. Fisherbrand™ Erlenmeyer Flask Clamps are manufactured using strong and durable stainless steel for superior stability.
24.00€ - 44.10€
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
10619474
|
Fisherbrand™
1007553 |
50 mL Erlenmeyer Flask | |||||||
10136372
|
Fisherbrand™
1007555 |
250mL Erlenmeyer flask |
30.75€
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
10084622
|
Fisherbrand™
1007557 |
500 mL Erlenmeyer Flask |
36.90€
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
10562963
|
Fisherbrand™
1007558 |
1000mL erlenmeyer flask |
44.10€
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
Description
- Safe and secure attachment
- Manufactured from strong and durable stainless steel.
- Provides superior stability
Specifications
Flask Clamp | |
Ultrasonic Cleaner |