Learn More
Fisherbrand™ Comb for use with FB-VE10-1 Vertical Electrophoresis System
For use with Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System.
Brand:  Fisherbrand™ MP-12C
41.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11953015
  • 42.30€ / Each
Only null left
Estimated Shipment: 05-11-2025
Specifications
Owl P8 Series Electrophoresis System
10 x 10 cm
0.08 cm
5.9 in.
15 cm
3.9 in.
13 cm
0.43 cm
13 cm
Comb
12
55 μL
3.9 in.
10 cm
0.16 in.
10 cm
5.11 in.