Learn More
Fisherbrand™ Vortex Mixer Accessories: Ampule Tube Holders
For use with Fisherbrand vortex mixers.
Brand:  Fisherbrand™ 80863005
100.82 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15716959
  • 123.00€ / Each
Only null left
Estimated Shipment: 20-10-2025
Add to basket
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Mixes storage vials and test tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15 to 17 mm dia.
Ampoule Holder
Vortex mixer