Learn More
Fisherbrand™ Microtube Holder for Heavy-duty Vortex Mixer
For use with heavy-duty vortex mixer.
Brand: Fisherbrand™ 80863046
37.13 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Holds a range of Micro-Tube sizes and numbers
- Requires Universal Holder
Specifications
0.5 mL | |
Microtube Holder | |
Foam insert | |
Universal holder |
0.5 mL | |
Heavy-duty vortex mixer | |
52 |