Learn More
BINDER™ Factory Calibration Certificate
For Use With: BINDER units C 170
Brand: BINDER™ 8012-0228
501.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Temperature measurement in center of chamber
- CO2 measurement performed using test gas at 37°C and 5% CO2
Specifications
Calibration certificate, temperature and CO2 - for temperature and CO2, temperature measurement in center of chamber, CO2 measurement performed using test gas at 37°C and 5 % CO2 | |
BINDER Chambers Series CB-S | |
Calibration Certificate | |
Temperature and Carbon Dioxide |