Learn More
Taylor Wharton™ Extracting Tubes
502.00€ - 590.00€
Specifications
Type | Extracting Tube |
---|---|
For Use With (Application) | Transfer of cryogenic fluids |
For Use With (Equipment) | 3/8 in. MNPT |
Product Code | Brand | Length (Metric) Exterior | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) Exterior | Price | Quantity & Availability | |||||
11738429
|
Taylor Wharton™
17009C65W |
1200 mm |
502.00€
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
|||||
11758419
|
Taylor Wharton™
16009C66W |
1800 mm |
590.00€
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||||
Specifications
Extracting Tube | |
Transfer of cryogenic fluids | |
3/8 in. MNPT |