Learn More
Eppendorf™ Heat Sealer Film and Foils
Create 100% hermetic seal, eliminate evaporation, and prevent cross-contamination
148.00€ - 208.00€
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11518663
![]() |
Eppendorf™
0030127870 |
Storage Film |
159.00€
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11508663
![]() |
Eppendorf™
0030127854 |
PCR Foil |
148.00€
Pack of 100 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
11833432
![]() |
Eppendorf™
0030127838 |
PCR Film |
208.00€
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
- Requires Eppendorf heat sealer (Cat. No. E5390 000 016)
- Transparent heat sealing film allows sample monitoring in the plates
- Aluminum coated, heat-sealing foils are available in peel-off or pierceable versions
- Foil stripper tool removes heat-sealed film and foils completely
- Certified free of DNA, DNase, RNase, and PCR inhibitors
- Autoclavable
Specifications
Autoclavable | |
Yes |