Learn More
Merck Millipore Petri Dishes and Petri-Pad™ Dishes
Available with or without absorbent pads
Brand:  Merck Millipore PD20047S5
298.44 EUR valid until 2025-06-21
Use promo code "24097" to get your promotional price.
 View more versions of this product
Product Code. 11993856
  • 534.00€ / Pack of 600
Outlet Corner
Use promo code 24098 at checkout
View Available Promotions
Only null left
Estimated Shipment: 23-06-2025
Add to basket
Requires : When using with liquid medium, add sterile absorbent pads (order Cat. No. AP10 045 S0, 47mm Sterile Pads; or AP10 045 S1, Millipore Pad Dispenser with Sterile Pads)
Description
Description
Available with or without absorbent pads
Petri Dishes
  • 47mm sterilized Petri dishes
  • Use with solid agar medium
  • Dishes are made of molded polystyrene
Petri-Pad Dishes
  • Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
  • Eliminate labor and cost of hand-loading pads and minimize contamination
  • Pads are pure cellulose