Learn More
Bel-Art™ Ellipso-Spoon and Spatula Samplers
Special curved shape prevents missed sample accumulation in corners.
Brand: Bel-Art™ H36807-0021
32.04 EUR valid until 2025-08-31
Use promo code "24124" to get your promotional price.
This item is not returnable.
View return policy
Description
- Crystals, powders, granulated and solid substances, and thick liquids retrieved
- Includes spoon end and spatula end
- Made of polished 304 grade stainless steel
Specifications
Spoon and Spatula Sampler | |
Autoclavable | |
15 x 35 mm bowl | |
Designed to extracts crystals, granulated and solid substances, thick liquids and delicate materials | |
21 cm |
Stainless Steel | |
10 mL Bowl | |
Polished | |
Bottles and containers |