Learn More
Ansell EDGE™ 14-662 Certified PVC-Coated Chemical Resistant Gloves - Safety Cuff
High quality gloves provide reliable protection to workers who are exposed to oil, grease, caustics, and alcohols. Textured finish protects against abrasions while also enhancing grip capabilities in wet and oily conditions.
54.70€
Specifications
Certifications/Compliance (Europe) | EN 407:2004, EN 420:2003 + A1:2009, REACH Compliant |
---|---|
Applications | Industrial |
Color | Blue |
Exterior Finish | Fully Textured |
Latex Free | Yes |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16679093
|
Ansell
14662080 |
8 |
54.70€
12 pairs |
Estimated Shipment: 24-11-2025
Log in to see stock. |
|||||
16689093
|
Ansell
14662090 |
9 |
54.70€
12 pairs |
In Stock
Log in to see stock. ![]() |
|||||
16699093
|
Ansell
14662100 |
10 |
54.70€
12 pairs |
Estimated Shipment: 23-09-2025
Log in to see stock. |
|||||
16609103
|
Ansell
14662110 |
11 |
54.70€
12 pairs |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
Description
- Protects workers that are exposed to oil, grease, acid, caustics and alcohols
- Rough finish protects against abrasions while enhancing grip, even in wet and oily conditions
- Flexibility and comfort leads to less hand fatigue and better worker efficiency
Specifications
EN 407:2004, EN 420:2003 + A1:2009, REACH Compliant | |
Blue | |
Yes | |
Chemical-resistant Gloves | |
Type C | |
Safety |
Industrial | |
Fully Textured | |
PVC | |
3121A | |
Yes | |
14-662 |