Learn More
Dioxane solution EU Pharmacopoeia, Fisher chemical™
BIOPHARMA
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
228.00€ - 238.00€
Specifications
Type Mass Spectrometry Standards
Grade Eu Ph Ref: 1032001
Chemical Name or Material Dioxan stock solution.
Products 2
Product Code Brand Quantity Packaging Purity Grade Notes Price Quantity & Availability  
Product Code Brand Quantity Packaging Purity Grade Notes Price Quantity & Availability  
12940404
BIOPHARMA
View Documents
Fisher Chemical
RP 066 100
100 mL Bottle Eu Ph Ref: 1032001
238.00€
100mL
12950404
BIOPHARMA
View Documents
Fisher Chemical
RP 067 100
100 mL Bottle Eu Ph Ref: 1032002
228.00€
100mL
Specifications
Mass Spectrometry Standards
Eu Ph Ref: 1032001
Dioxan stock solution.
Videos
Safety and Handling
Safety and Handling
H225; H304; H315; H336; H410
P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501
Danger
missing translation for 'shelfLife' : 12 months
Product Identifier
  • Dioxan stock solution. European Pharmacopoeia Ref: 1032001
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,4-dioxane