Learn More
Thermo Scientific™ Culti-Loops™ Klebsiella pneumoniae subsp. pneumoniae ATCC™ 35657™
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
ATCC® | 35657TM* |
Description | Klebsiella pneumoniae subsp. pneumoniae |
Detects | Klebsiella pneumoniae subsp. Pneumoniae |
Product Type | Culti-Loops Quality Control Organism |
Target Organism Class | Gram-negative Rods |
Test Type | Qualitative QC |
Quantity | 5 Loops/Pack |
Strain Designations | LRA 24 11 73 |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.