Learn More
Sartorius Cubis™ II Precision (2-place) Balance, MCE User Interface
Cubis™ II Precision (2-Place) Balance features fully customizable hardware, software, and connectivity
5205.00€ - 14735.00€
Specifications
Connection Type | 2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port |
---|---|
Display Type | TFT Touchscreen |
Model | No draft shield |
Type | Precision Balance |
Warranty | 10 years |
Product Code | Brand | Capacity | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Includes | Price | Quantity & Availability | ||||
16477445
![]() ![]() |
Sartorius
MCE6202P-2S00-0 |
1.5 kg, 3 kg, 6.2 kg | Balance, Accessories, Power supply, Manual, Factory test report |
5205.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16223272
![]() ![]() |
Sartorius
MCE32202P0-S00 |
4.2 kg, 32.2 kg | Balance, Accessories, Power supply, Manual, Factory test report, QP99 software |
14735.00€
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16203282
![]() |
Sartorius
MCE4202S0-S00 |
4.2 kg | Balance, Accessories, Power supply, Manual, Factory test report, QP99 software |
5695.00€
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16223282
![]() ![]() |
Sartorius
MCE6202S-2S00-0 |
6.2 kg | Balance, Accessories, Power supply, Manual, Factory test report, QP99 software |
6680.00€
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16407495
![]() ![]() |
Sartorius
MCE8202S-2S00-0 |
8.2 kg | Balance, Accessories, Power supply, Manual, Factory test report |
7475.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16203262
![]() |
Sartorius
MCE2202S0-S00 |
2.2 kg | Balance, Accessories, Power supply, Manual, Factory test report, QP99 software |
5370.00€
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Monolithical weighing system
- Integrated climate sensors
- Individual sample holders
- Error-free operation
- Individual QApp workflows
- Motorized auto-leveling
- Full end-to-end data integrity
- Integrated audit-trail
- State-of-the-art user management
- Excellent service support
- Integrated status center
- Integrated service functions
- Preventive maintenance on accredited standards
- TFT touch screen operation with readable display and easy to operate user interface
- Software Factory installed basic set of essential weighing applications
- Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Warranty
10-year warranty
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port | |
No draft shield | |
10 years | |
50/60 Hz (± 5%) |
TFT Touchscreen | |
Precision Balance | |
100/240 V | |
Class II |