Learn More
Tenak™ Green Cardboard Cryobox
Shop All TENAK Products
Click to view available options
Dimensions (L x W x H):
133 x 133 x 32 mm
133 x 133 x 50 mm
133 x 133 x 75 mm
136 x 136 x 32 mm
136 x 136 x 50 mm
133 x 133 x 100 mm
133 x 133 x 130 mm
136 x 136 x 100 mm
136 x 136 x 130 mm
Unit Size:
Pack of 36
Description
- Constructed of fibreboard/cardboard
- Water-repellent for longer durability
- Can be used with a large range of grid dividers
Specifications
Specifications
Color | Green |
Material | Cardboard |
Shape | Square |
Type | Cryo Box |
Dimensions (L x W x H) | 136 x 136 x 50 mm |