Learn More
DuPont™ Tyvek 200 Easysafe Hooded Coverall
Offering adequate Type 5 and 6 barrier protection and remains both air and moisture vapour permeable. DuPont™ Tyvek™ 200 Easysafe Hooded Coveralls are made from a new and unique optimized polyethylene nonwoven that is softer with a more open structure than Tyvek™ 500 and 600.
Brand: DuPont™ D14886064
4.89 EUR valid until 2025-09-30
Use promo code "24380" to get your promotional price.
Description
- Robust yet lightweight
- Elasticated face, wrists, waist and ankles
- Antistatic treatment on the outside
- Stitched external seams in yellow for visual identification and differentiation
Specifications
Hooded Coveralls | |
Polyethylene | |
Elastic | |
2-panel | |
Stitched | |
(EU) 2016/425, EN 1073-2, EN 1149-1 |
X-Large | |
White | |
Hooded | |
Zipper | |
Category III, Type 5 and 6 | |
108 to 116 cm |