Learn More
SNV Comines™ Cotton Laboratory Coats
Provides general light protection against splashes in the laboratory. SNV Comines™ Cotton Laboratory Coats are available in a range of sizes.
Brand:  SNV Comines™ ANTLB00400T0
17.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12605306
  • 25.00€ / Each
Only null left
Estimated Shipment: 03-09-2025
Description
  • 4 buttons
  • Collared
Specifications
Specifications
Lab Coat
Women's
White
4
38
Cotton
Buttons
84 cm