Learn More
Corning™ Freezing Cartridges for CoolBox™ 30 Cooling System
Click to view available options
Color:
Green
Unit Size:
Pack of 3
Description
Limited quantities remain. Buy now at a deep discount.
Freezing Cartridge for CoolBox, green, 3 per pack
Specifications
Specifications
Type | Cooling Cartridge |
Color | Green |
No. per Pack | 1/Pk. |
No. per Case | 3/Cs. |
For Use With (Application) | Freezing |
For Use With (Equipment) | CoolBox 30 Cooling System |
Dimensions (L x W x H) | 10.8 x 10.2 x 2.5 cm (4.3 x 4.0 x 1.0 in.)p |