Learn More
Agilent BioTek™ Confocal Imaging Accessory Kit
For use with Cytation C10 Confocal Imaging Reader.
Brand: Agilent BioTek™ 1940027
This item is not returnable.
View return policy
Description
- For use with Cytation C10 Confocal Imaging Reader.
- Designed to provide the essential objectives, filters cubes, LEDs and adapters required for use across a broad range of confocal imaging workflows.
- Includes protective case for convenience and ease of use.
Specifications
Accessory Kit | |
Cytation C10 Confocal Imaging Reader | |
Non-sterile |