Learn More
Tenak™ Comfort Rack for Upright Freezers, 85 mm Box Height
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Brand: Tenak™ TE24334
193.58 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
Specifications
85 mm (Box) | |
Upright Freezers | |
4 x 4 |
562 x 140 x 348 mm | |
Boxes (16) | |
Stainless Steel |