Learn More
Copan Italia™ Combined Loop and Handle
71.00€
Specifications
Product Type | Combined Loop and Handle |
---|---|
Gauge | 26 G |
Quantity | 5 Pk. |
Calibrated | No |
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
11706528
|
Copan Italia™
316C05T |
2 mm |
71.00€
Pack of 5 |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
11726528
|
Copan Italia™
318C05T |
4 mm |
71.00€
Pack of 5 |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
11736528
|
Copan Italia™
319C05T |
5 mm |
71.00€
Pack of 5 |
Estimated Shipment: 10-10-2025
Log in to see stock. |
|||||
11716528
|
Copan Italia™
317C05T |
3 mm |
71.00€
Pack of 5 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
Specifications
Combined Loop and Handle | |
26 G | |
5 Pk. | |
No |