Learn More
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in.
StopCock spigots, ball valves and other accessories can be connected to most commercially available containers, with the LaboPlast™ thread adapters.
Brand: Buerkle™ 0600-0010
6.80 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- All adapters have a standard outer thread R 3/4 in.
Specifications
Polypropylene | |
Hunersdorff External, 3/4 in. Internal | |
Red | |
Thread Adapter |