Learn More
Memmert™ Clip for 300 mL Flask
Clip for 300 mL flask (14 flasks max.)
Brand:  Memmert™ WTB50B02694
29.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17110395
  • 30.75€ / Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
Description
Description
  • Accessory
  • Max. flasks see sketch on demand
Specifications
Specifications
WTB50
Clip for 300 mL flask