Learn More
Fisherbrand™ Clear Graduated Glass Bottles with White Polypropylene Caps
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
120 mL
240 mL
480 mL
Quantity:
24
48
96
144
Specifications
Specifications
Capacity (Metric) | 240 mL |
Capacity (English) | 8 oz. |
Color | Clear |
Diameter (Metric) Outer Neck | 54 mm |
Diameter (Metric) Inner Neck | 40 mm |
Height (Metric) With Closure | 130 mm |
Material | Glass (Type 3) |
Quantity | 96 |
Closure Color | White |
Neck Finish | 58-400 |