Learn More
Fisherbrand™ Class B Glass Tall Form Measuring Cylinders
Measure and pour liquids with precision with these cylinders. Fisherbrand™ Class B Glass Tall Form Measuring Cylinders are manufactured from calibrated, clear borosilicate glass.
Brand:  Fisherbrand™ DBG504
4.57 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11807501
  • 15.30€ / Each
Only null left
Estimated Shipment: 09-09-2025
Description
  • Graduated
  • Stable hexagonal foot
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
ISO 4788
195 mm
Graduated Measuring Cylinder
26 mm
1 mL
±1mL
B
±1 mL
Borosilicate Glass
50 mL
Autoclavable
500°C