Learn More
Hirschmann™ Class A Measuring Cylinders, Blue Graduation
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Blue Graduation are manufactured from durable DURAN™ glass tubing.
26.15€ - 149.00€
Specifications
Base Type | Hexagonal |
---|---|
Certifications/Compliance | DIN EN ISO 4788 |
Class | A |
Type | Graduated Measuring Cylinder |
Scale Color | Blue |
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
11786482
|
Hirschmann™
2240154 |
5 mL | Certified Conformity |
28.00€
Pack of 2 |
In Stock
Log in to see stock. |
||||
11748041
|
Hirschmann™
2240160 |
10 mL | Certified Conformity |
26.15€
Pack of 2 |
In Stock
Log in to see stock. |
||||
11758041
|
Hirschmann™
2240170 |
25 mL | Certified Conformity |
27.60€
Pack of 2 |
In Stock
Log in to see stock. |
||||
11768041
|
Hirschmann™
2240175 |
50 mL | Certified Conformity |
29.05€
Pack of 2 |
In Stock
Log in to see stock. ![]() |
||||
11778041
|
Hirschmann™
2240180 |
100 mL | Certified Conformity |
31.10€
Pack of 2 |
In Stock
Log in to see stock. ![]() |
||||
11788041
|
Hirschmann™
2240186 |
250 mL | Certified Conformity |
59.35€
Pack of 2 |
In Stock
Log in to see stock. ![]() |
||||
11798041
|
Hirschmann™
2240190 |
500 mL | Certified Conformity |
99.05€
Pack of 2 |
In Stock
Log in to see stock. ![]() |
||||
10569242
|
Hirschmann™
2240193 |
1000 mL | Certified Conformity |
76.60€
Each |
In Stock
Log in to see stock. ![]() |
||||
10440261
|
Hirschmann™
2240194 |
2000 mL | Certified Conformity |
137.00€
Each |
In Stock
Log in to see stock. ![]() |
||||
10312743
|
Hirschmann™
224016027 |
10 mL | USP |
27.80€
Pack of 2 |
In Stock
Log in to see stock. |
||||
10747793
|
Hirschmann™
224017027 |
25 mL | USP |
29.35€
Pack of 2 |
In Stock
Log in to see stock. |
||||
10097262
|
Hirschmann™
224017527 |
50 mL | USP |
31.00€
Pack of 2 |
In Stock
Log in to see stock. |
||||
10597193
|
Hirschmann™
224018027 |
100 mL | USP |
32.90€
Pack of 2 |
In Stock
Log in to see stock. |
||||
10765683
|
Hirschmann™
224018627 |
250 mL | USP |
63.25€
Pack of 2 |
In Stock
Log in to see stock. |
||||
10436973
|
Hirschmann™
224019027 |
500 mL | USP |
106.00€
Pack of 2 |
Estimated Shipment: 25-07-2025
Log in to see stock. |
||||
10241253
|
Hirschmann™
224019327 |
1000 mL | USP |
81.50€
Each |
In Stock
Log in to see stock. |
||||
10332933
|
Hirschmann™
224019427 |
2000 mL | USP |
149.00€
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
Description
- With spout, main point ring graduation
- Blue graduation
- Conformity certified, with dated batch identification
- Acc. to DIN EN ISO 4788
- Corrosion-free and resistant to thermal shock
Specifications
Hexagonal | |
A | |
Blue | |
Glass |
DIN EN ISO 4788 | |
Graduated Measuring Cylinder | |
Round |