Learn More
Sartorius Stedim Biotech™ Chromatography Papers, Sheets, Grade FN 8
Shop All Sartorius Products
Click to view available options
Unit Size:
Pack of 50
Description
- Highly pure paper grade.
- Ideal for blotting and chromatography.
- Ideal for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Non-sterile, bulk packed.
Specifications
Specifications
| Description | 50/Pk, 280 g/m2 |
| Dimensions (L x W) | 470 mm × 580 mm |
| Film Thickness | 0.9 mm |
| Format | Sheet |
| Grade | FN 8 |
| Material | Cotton Linters (> 98 % alpha-cellulose; < 0.04 % ash) |