Learn More
Chroma Gesellschaft™ Hand Operated Crimper
289.00€
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
12423030
|
JG Finneran Associates
9300-20 |
20 mm |
289.00€
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
|||||
10710862
|
JG Finneran Associates
9300-11 |
11 mm |
289.00€
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
10597542
|
JG Finneran Associates
9300-08 |
8 mm |
289.00€
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
|||||