Learn More
Swann-Morton™ Carbon Steel Sterile Scalpel Blade
Brand:  Swann-Morton™ 0212
25.69 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11882431
  • 35.95€ / Pack of 100
Only null left
Estimated Shipment: 16-09-2025
Description
Order Info
Size
Specifications
Specifications
Carbon Steel
# 25
Surgeon Grade
10/Pk.
Scalpel Blade
Fits # 4, # 4L, # 4 Graduated and # 6B Handles