Learn More
Carbenicillin disodium salt
Carbenicillin disodium salt, CAS # 4800-94-6, is the disodium salt form of carbenicillin, a semisynthetic compound with broad-spectrum antibacterial and β-lactamase-resistant activities. | CAS: 4800-94-6 | C17H16N2Na2O6S | 422.36 g/mol
253.00€
Chemical Identifiers
CAS | 4800-94-6 |
---|---|
Molecular Formula | C17H16N2Na2O6S |
Molecular Weight (g/mol) | 422.36 |
MDL Number | MFCD00077683 |
InChI Key | RTYJTGSCYUUYAL-YCAHSCEMSA-L |
PubChem CID | 20933 |
ChEBI | CHEBI:34609 |
SMILES | [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
Description
Carbenicillin disodium salt is a water-soluble antibiotic which is effective against gram-negative bacteria.Carbenicillin disodium salt is widely utilized as an antibiotic which is used to control bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. It has been involved in the study of the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Carbenicillin disodium salt is a semisynthetic compound with antibacterial and β-lactamase-resistant activities and is effective against gram-negative bacteria
- In in vitro studies, carbenicillin disodium salt can totally suppress the proliferation of Agrobacterium
Application
- Due to its antibacterial activity, this compound can be used to control bacterial cell wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. This can lead to incomplete bacterial cell wall synthesis to eventually cause cell lysis
- It has also been used in studies to evaluate the role of penicillin-sensitive transpeptidases in cell wall biosynthesis
Chemical Identifiers
4800-94-6 | |
422.36 | |
RTYJTGSCYUUYAL-YCAHSCEMSA-L | |
CHEBI:34609 |
C17H16N2Na2O6S | |
MFCD00077683 | |
20933 | |
[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
Specifications
Carbenicillin Disodium Salt | |
4800-94-6 | |
C17H16N2Na2O6S | |
5722128 | |
Soluble in water and alcohol. | |
[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O | |
422.36 | |
CHEBI:34609 | |
Carbenicillin disodium salt |
Powder | |
White | |
MFCD00077683 | |
14,1792 | |
RTYJTGSCYUUYAL-YCAHSCEMSA-L | |
disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
20933 | |
422.36 |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P271-P272-P280g-P285-P302+P352-P304+P340-P333+P313-P342+P311-P363-P501c
H317-H334-H335
missing translation for 'einecsNumber' : 225-360-8
missing translation for 'rtecsNumber' : ON9105000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
- Carbenicillin disodium salt
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Disodium [2S-(2.alpha.,5.alpha.,6.beta.)]-6-(carboxylatophenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.