Learn More
Erlab™ Captair Smart Glass Work Surface with Retention Tray
Erlab™ Captair Smart Work Surface with Retention Tray is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
Brand: Erlab™ I0161020001
510.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Tempered enameled glass worktop
- Includes retention tray filtering fume hoods
- Jointed frame
- Ergonomic arm rest for a comfortable working position
Specifications
Smart 391 Fume Hood | |
470 mm | |
903 mm | |
Tempered Glass |
Retention Tray | |
Jointed Frame | |
Work Surface |