missing translation for 'onlineSavingsMsg'
Learn More

Calix[8]arene, 97%, Thermo Scientific Chemicals

Product Code. 11400247
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 82452-93-5 | C56H48O8 | 848.992 g/mol

It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.

Solubility
Sparingly soluble in water (5.8E-6 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82452-93-5
Molecular Formula C56H48O8
Molecular Weight (g/mol) 848.992
MDL Number MFCD00143914
InChI Key HDPRHRZFFPXZIL-UHFFFAOYSA-N
Synonym calix 8 arene, calix 8 arene;, p-h-calix 8 arene, octahydroxycalix 8 arene, para-h-calix 8 arene;, calix 8 arene, technical ch, 49,50,51,52,53,54,55,56-octahydroxycalix 8 arene, 49,50,51,52,53,54,55,56-octahydroxycalix 8 arene;, 49,50,51,52,53,54,55,56-octahydroxycalix 8-arene;, 49,50,51,52,53,54,55,56-octakis hydroxy calix 8 arene;
PubChem CID 2724886
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=C(C(=CC=C4)CC5=C(C(=CC=C5)CC6=CC=CC(=C6O)CC7=CC=CC(=C7O)CC8=CC=CC(=C8O)CC9=CC=CC1=C9O)O)O)O)O

Specifications

Melting Point >450°C
Quantity 5 g
Solubility Information Sparingly soluble in water (5.8E-6g/L) (25°C).
Formula Weight 848.99
Percent Purity 97%
Chemical Name or Material Calix[8]arene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.