Learn More
BINDER™ Calibration Certificate, Temperature
Calibration Certificate for temperature, measurement in center of chamber at specified temperature.
Brand:  BINDER™ 8012-1608
 View more versions of this product
Product Code. 15913012
  • 1915.00€ / Each
Description
  • Measurement at specified temperature
  • Including certificate
  • Various points of measurement
Specifications
Specifications
BINDER Chambers (Models MKFT 115, MKFT 240, MKFT 720)
27 Measurement Points
Calibration Certificate (Temperature)
Calibration certificate, temperature - temperature measurement incl. certificate and 27 measuring points (at specified temperature)
60 to 120 min.
Temperature Measurement