Learn More
Cadence Science™ Deflected Noncoring Septum Penetration Needles
Click to view available options
Needle Gauge:
18 G
20 G
Length (English) Needle:
6 in.
8 in.
12 in.
Unit Size:
Pack of 12
Description
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
- Penetration needles with deflected noncoring septum
- For noncoring penetration of septa and rubber closures involving air-sensitive and pyrophoric compounds
Specifications
Specifications
Material | Nickel-plated Brass (Hub), Stainless Steel (Tube) |
Needle Gauge | 20 G |
For Use With (Application) | Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures |
Includes | Deflected non-coring septum |
Length (English) Needle | 8 in. |
Product Type | Penetration Needle |
Quantity | 12 Needles |
Needle Point Style | Deflected End |