Learn More
BUCHI Glass Columns C-690
Shop All Buchi Products
Click to view available options
Length (Metric):
460 mm
Diameter (Metric):
26 mm
Max. Pressure:
580 psi
Unit Size:
Each
Specifications
Specifications
Diameter (Metric) | 26 mm |
For Use With | Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators |
Length (Metric) | 460 mm |
Max. Pressure | 580 psi |
Material | Borosilicate Glass |
Type | Glass Column |