Learn More
BUCHI Drying Flasks for Rotary Evaporators
Special shaped flasks for powder drying
Brand:  BUCHI 000453
200.94 EUR valid until 2025-12-20
Use promo code "24121" to get your promotional price.
 View more versions of this product
Product Code. 10729782
  • 217.11€ / Each
Equipment End of Year Sale
Use promo code 24121 at checkout
View Available Promotions
Only null left
Estimated Shipment: 04-09-2025
Description
  • For use with the Rotavapor R 3 and R-210/R-215
  • Available in multiple sizes with different standard joints
Specifications
Specifications
CE
Powder Drying Flask
Plastic and Glass
29.2/32
1000 mL
Standard Glass
Rotary evaporator
29.2/32
60°C