Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Beaker Positioning Covers
For use with Branson ultrasonic cleaning baths
Brand:  Branson Ultrasonics™ 100-032-520
41.60 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15338634
  • 41.89€ / Each
Only null left
Estimated Shipment: 18-09-2025
Description
For use with Branson ultrasonic cleaning baths
  • Helps positon beakers securely in bath
Specifications
Specifications
Beaker Positioning Cover
Beaker in Ultrasonic Benchtop Cleaning Bath, 2 x 600 mL, Fits Model 3800
Helps position beakers securely in bath