Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Perforated Trays
For use with Branson ultrasonic baths
Brand:  Branson Ultrasonics™ 100-410-162
133.04 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10644175
  • 172.00€ / Each
Only null left
Estimated Shipment: 22-09-2025
Description
For use with Branson ultrasonic baths
  • Accessories for ultrasonic bench-top system
Specifications
Specifications
Perforated Tray
0.75 gal.
Stainless Steel
3.4 L
Ultrasonic Bath, Model 2800