Learn More
BRAND™ Replacement Dispenser Cartridges for seripettor™ and seripettor™ pro
Replacement Cartridges for seripettor™ and seripettor™ pro, and QuickSip™ bottletop dispensers
Brand: BRAND™ 704506
73.15 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sterile or non-sterile
- For use with seripettor™, seripettor™ pro and QuickSip™ bottletop dispensers
Specifications
Not Autoclavable | |
Seripettor / Seripettor pro / QuikSip | |
Sterile |
10 mL | |
7/Pk. | |
10/Pk. |